Learn More
Thermo Scientific™ HyperSep™ Florisil SPE 96-Well Plates and Individual Wells
Catalog No. 03251227 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Magnesia-loaded Silica Gel
Catalog No. Sorbent
03-251-227 HyperSep Magnesia-loaded Silica Gel
1 options
Catalog No. 03-251-227 Supplier Thermo Scientific™ Supplier No. 60300723
Isolate polar compounds from nonpolar matrices with these Florisil SPE well plates, which are especially well-suited for extraction of chlorinated pesticides.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep Florisil SPE 96-Well Plates and Individual Wells are ideal for the isolation of polar compounds from nonpolar matrices. Typical applications include:
Extraction of pesticides using official methods
Extraction of polychlorinated biphenyls (PCBs) in transformer oil
Alcohol, aldehydes, amines and drugs
Extremely polar in nature, HyperSep Florisil SPE 96-Well Plates and Individual Wells are especially well-suited for extraction of chlorinated pesticides. The magnesia-loaded silica gel offers rapid flow rates for large sample volumes.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Description Individual SPE Wells; Bed weight 50 mg
For Use With (Application) Isolation of Polar Compounds From Non-Polar Matrices
Particle Size 40 to 60 μm
Sorbent HyperSep Magnesia-loaded Silica Gel
Type Individual Well