Learn More
Thermo Scientific™ HyperSep™ Florisil Plates
Catalog No. 03251230 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Florisil 10 mg/1 mL 96 Removable Well Plate, 1 pk
Florisil 100 mg/1 mL 96 Removable Well Plate, 1 pk
Florisil 25 mg/1 mL 96 Removable Well Plate, 1 pk
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
03-251-230 Florisil 25 mg/1 mL 96 Removable Well Plate, 1 pk
03-251-229 Florisil 10 mg/1 mL 96 Removable Well Plate, 1 pk
03-251-232 Florisil 100 mg/1 mL 96 Removable Well Plate, 1 pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-251-230 Supplier Thermo Scientific™ Supplier No. 60300726
Isolate polar compounds from nonpolar matrices with these Florisil SPE well plates, which are especially well-suited for extraction of chlorinated pesticides.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ Florisil Plates are ideal for the isolation of polar compounds from nonpolar matrices. Typical applications include:
  • Extraction of pesticides using official methods
  • Extraction of polychlorinated biphenyls (PCBs) in transformer oil
  • Alcohol, aldehydes, amines and drugs
Extremely polar in nature, HyperSep Florisil SPE 96-Well Plates and Individual Wells are especially well-suited for extraction of chlorinated pesticides. The magnesia-loaded silica gel offers rapid flow rates for large sample volumes.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Description Florisil 25 mg/1 mL 96 Removable Well Plate, 1 pk
Bed Weight 25 mg
For Use With (Application) Isolation of Polar Compounds From Non-Polar Matrices
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep Magnesia-loaded Silica Gel
Type 96-well Plate
Volume (Metric) Well 1 mL