Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypersep™ Hypercarb™ Fixed Well Plate

Catalog No. 13622468
Change view
Click to view available options
Bed Weight:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
13-622-468 25 mg
60-306-503 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-622-468 Supplier Thermo Scientific™ Supplier No. 60306502
Add to Cart
Add to Cart

Unique material for retention of highly polar compounds.

Thermo Scientific™ HyperSep™ and Hypercarb™ SPE contains flat,100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents

  • pH stable 0 to 14
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • Interaction mechanism with polar molecules
TRUSTED_SUSTAINABILITY

Specifications

Bed Weight 25 mg
For Use With (Equipment) Universal Vacuum Manifold
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HypeSep Hypercarb
Type 96-well Plate
Quantity 1/Ea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.