Learn More
Thermo Scientific™ HyperSep™ Phenyl SPE 96-Well Plates and Individual Wells
Catalog No. 03251217 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Phenyl
Catalog No. Sorbent
03-251-217 HyperSep Phenyl
1 options
Catalog No. 03-251-217 Supplier Thermo Scientific™ Supplier No. 60300681
Obtain improved extraction of aromatic compounds and basic analytes with these phenyl SPE well plates offering alternative selectivity for retention of basic compounds.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
In Thermo Scientific™ HyperSep Phenyl SPE 96-Well plates and Individual Wells, the electron density of the aromatic ring provides different selectivity than other nonpolar sorbents for retention of basic compounds. The cartridges are commonly used for nonpolar interactions, such as:
Extraction of aromatic compounds
Extraction of basic analytes
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Description Individual SPE Wells; Bed weight 10 mg
For Use With (Application) Extraction of Aromatic Compounds and Basic Analytes
Particle Size 40 to 60 μm
Sorbent HyperSep Phenyl
Type Individual Well