Learn More
Thermo Scientific™ HyperSep™ Pressure Manifold Waste Container
Catalog No. 60104270 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Positive Pressure Manifold
Catalog No. For Use With (Equipment)
60-104-270 Positive Pressure Manifold
1 options
Catalog No. 60-104-270 Supplier Thermo Scientific™ Supplier No. 60104270
Add to Cart
Add to Cart
Collect waste from the Thermo Scientific™ HyperSep™ Positive Pressure Manifold using this pre-drilled waste container.
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Waste Container to collect waste from the manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
TRUSTED_SUSTAINABILITY
Specifications
Description Pre-Drilled Waste Container, 1 pk
For Use With (Application) SPE
For Use With (Equipment) Positive Pressure Manifold
Product Line HyperSep
Type Pre-Drilled Waste Container