Learn More
Thermo Scientific™ HyperSep™ Protein Precipitation Plate
Catalog No. 60304201 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
Protein Denaturing in Biological Samples
Catalog No. For Use With (Application)
60-304-201 Protein Denaturing in Biological Samples
1 options
Catalog No. 60-304-201 Supplier Thermo Scientific™ Supplier No. 60304201
Add to Cart
Add to Cart
Get a quick, effective approach for removal of proteins from biological compounds using the protein crash technique with these 96-well protein precipitation plates.
Thermo Scientific™ HyperSep™ Protein Precipitation Plates provide a quick, effective approach for removal of proteins from biological compounds using the protein crash technique. In combination with SPE and SLE, the Protein Precipitation Plate offers a comprehensive range of options for sample preparation of biological based compounds. The 96-well plates are ideal for use in automated, high throughput systems.
HyperSep Protein Precipitation Plates offer these features:
  • Dual frit design
  • Hydrophobic/oleophobic frits enable only precipitation of proteins
  • Pore size is optimized for ideal flow rate
  • Specially selected polypropylene for low extractables
TRUSTED_SUSTAINABILITY
Specifications
Description Protein Precipitation Plate
For Use With (Application) Protein Denaturing in Biological Samples
Product Line HyperSep
Sorbent HyperSep Protein
Type Protein Precipitation Plate
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.