Learn More
Thermo Scientific™ HyperSep™ Retain AX SPE 96-Well Plates and Individual Wells
Shop All Thermo Scientific ProductsDescription
Achieve superior recoveries for acidic analytes with Thermo Scientific™ HyperSep™ Retain AX SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with quaternary amine functional groups. Their use in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
- High, consistent recoveries
- Ideal for large samples
- Fast and easy sample preparation and method development
- Reproducible, reliable sample preparation in a traditional format
- pH stable 0 to 14
- Available in a wide range of chemistries and bed weights
Specifications
Specifications
| Description | Individual SPE Wells; Bed weight 60 mg |
| For Use With (Application) | Analysis of THC and its Metabolites |
| Particle Size | 30 to 50 μm |
| Sorbent | HyperSep Retain AX |
| Type | Individual Well |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.