Learn More
Thermo Scientific™ HyperSep™ Retain CX SPE 96-Well Plates and Individual Wells
Shop All Thermo Scientific ProductsDescription
Achieve consistent, high recoveries for basic compounds with these Thermo Scientific™ HyperSep™ Retain™ CX SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with sulfonic acid functional groups to give superior recoveries for basic analytes. Used in SPE sample preparation, these cation exchange well plates and individual wells can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
- High, consistent recoveries
- Ideal for large samples
- Fast and easy sample preparation and method development
- Reproducible, reliable sample preparation in a traditional format
- pH stable 0 to 14
- Available in a wide range of chemistries and bed weights
Specifications
Specifications
| Description | Individual SPE Wells; Bed weight 60 mg |
| For Use With (Application) | Analysis of Drugs of Abuse |
| Particle Size | 30 to 50 μm |
| Sorbent | HyperSep Retain CX |
| Type | Individual Well |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.