Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ Retain CX SPE 96-Well Plates and Individual Wells

Catalog No. 03251502 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Retain CX
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sorbent
03-251-502 HyperSep Retain CX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-251-502 Supplier Thermo Scientific™ Supplier No. 60303306

Achieve consistent, high recoveries for basic compounds with these Thermo Scientific™ HyperSep™ Retain™ CX SPE 96-Well Plates and Individual Wells.

Achieve consistent, high recoveries for basic compounds with these Thermo Scientific™ HyperSep™ Retain™ CX SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with sulfonic acid functional groups to give superior recoveries for basic analytes. Used in SPE sample preparation, these cation exchange well plates and individual wells can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Widely used in forensic and clinical analysis of a wide range of drugs of abuse, including basic and neutral drugs. Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Ideal for large samples
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • pH stable 0 to 14
  • Available in a wide range of chemistries and bed weights
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY

Specifications

Description 96-well plate; Bed weight 10 mg
For Use With (Application) Analysis of Drugs of Abuse
Particle Size 30 to 50 μm
Sorbent HyperSep Retain CX
Type 96-well Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.