Learn More
Thermo Scientific™ HyperSep™ Retain PEP Cartridges
Description
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Ideal for large samples, Hypersep Retain SPE cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
- High, consistent recoveries
- Fast and easy sample preparation and method development
- Reproducible, reliable sample preparation in a traditional format
- Available in a wide range of chemistries and bed weights
- Drugs and metabolites in biological fluids
- Peptides in serum, plasma or biological fluids
- Environmental samples
Specifications
Specifications
| Column Capacity (Metric) | 25 mL |
| Description | 1 g bed weight; 25 mL column volume |
| Bed Weight | 1000 mg |
| For Use With (Application) | Retention of Polar and Nonpolar Analytes |
| For Use With (Equipment) | Thermo Scientific HyperSep-96 System |
| Particle Size | 40 to 60 μm |
| Product Line | HyperSep |
| Sorbent | HyperSep Urea-Modified Polystyrene DVB |
| Type | Solid Phase Extraction Cartridge |
| Volume (Metric) | 25 mL |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.