Learn More
Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells
Shop All Thermo Scientific ProductsDescription
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
- [bullet] High, consistent recoveries
- [bullet] Fast and easy sample preparation and method development
- [bullet] Reproducible, reliable sample preparation in a traditional format
- [bullet] Available in a wide range of chemistries and bed weights
- [bullet] Drugs and metabolites in biological fluids
- [bullet] Peptides in serum, plasma or biological fluids
- [bullet] Environmental samples
Specifications
Specifications
| Description | Item Description |
| For Use With (Application) | Screening of Drugs, Metabolites, Peptides in Biological Samples |
| Particle Size | 30 to 50 μm |
| Sorbent | HyperSep Retain PEP |
| Type | Individual Well |