Learn More
Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells
Catalog No. 03251491 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Retain PEP
Catalog No. Sorbent
03-251-491 HyperSep Retain PEP
1 options
Catalog No. 03-251-491 Supplier Thermo Scientific™ Supplier No. 60303203
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells.
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features: Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • [bullet] High, consistent recoveries
  • [bullet] Fast and easy sample preparation and method development
  • [bullet] Reproducible, reliable sample preparation in a traditional format
  • [bullet] Available in a wide range of chemistries and bed weights
Applications:
  • [bullet] Drugs and metabolites in biological fluids
  • [bullet] Peptides in serum, plasma or biological fluids
  • [bullet] Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY
Specifications
Description Individual SPE Wells; Bed weight 30 mg
For Use With (Application) Screening of Drugs, Metabolites, Peptides in Biological Samples
Particle Size 30 to 50 μm
Sorbent HyperSep Retain PEP
Type Individual Well