Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ Retain PEP Plates

Catalog No. 60306208 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Weight:
30 mg
60 mg
Volume (Metric):
1 mL
2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Volume (Metric)
60-306-208 60 mg 2 mL
03-251-495 30 mg 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-306-208 Supplier Thermo Scientific™ Supplier No. 60306208
Get an Extra Case of Product with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Get balanced retention for polar and nonpolar analytes with HyperSep Retain PEP Plates.

Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Plates, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Features: Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • [bullet] High, consistent recoveries
  • [bullet] Fast and easy sample preparation and method development
  • [bullet] Reproducible, reliable sample preparation in a traditional format
  • [bullet] Available in a wide range of chemistries and bed weights
Applications:
  • [bullet] Drugs and metabolites in biological fluids
  • [bullet] Peptides in serum, plasma or biological fluids
  • [bullet] Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY

Specifications

Description Retain PEP 60 mg/2 mL 96 Fixed Well Plate, 1 pk
Bed Weight 60 mg
For Use With (Application) Retention of Polar and Nonpolar Analytes
Particle Size 30 to 50 μm
Product Line HyperSep
Sorbent HyperSep Urea-Modified Polystyrene DVB
Type Solid Phase Extraction Well Plate
Volume (Metric) 2 mL
Volume (Metric) Well 1 mL
Quantity 1 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.