Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ SAX Plates

Catalog No. 60307243 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
96 Fixed Well Plate
96 Removable Well Plate
Bed Weight:
10 mg
25 mg
50 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Bed Weight
60-307-243 96 Fixed Well Plate 50 mg
03-251-206 96 Removable Well Plate 50 mg
60-307-242 96 Fixed Well Plate 25 mg
03-251-204 96 Removable Well Plate 10 mg
03-251-205 96 Removable Well Plate 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-307-243 Supplier Thermo Scientific™ Supplier No. 60307243
Get an Extra Case of Product with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obtain excellent performance for extraction of carboxylic and other weak acids with these strong anion exchanger SPE well plates and individual wells.

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Thermo Scientific™ HyperSep™ SAX Plates feature a strong anion exchange sorbent for extraction of weak acids. Typical applications include:

  • Isolation of anionic proteins
  • Removal of acidic food pigments
  • Isolation of phenolic compounds
  • Nucleic acids, nucleotides and surfactants

HyperSep SAX Strong Anion Exchanger SPE 96-Well Plates and Individual Wells are ideally suited for the extraction of carboxylic and other weak acids. The chemistry extracts negatively charged compounds from both aqueous and nonaqueous solutions.
  • Amine group masks the effect of the carbon chain of the functional group
  • Selectivity can be tuned by choice of buffer

HyperSep SPE Cartridges and Plates

Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

TRUSTED_SUSTAINABILITY

Specifications

Description 96 Fixed Well Plate
Bed Weight 50 mg
For Use With (Application) Extraction of Weak Acids
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep SAX
Type 96-well Plate
Volume (Metric) 2 mL
Volume (Metric) Well 1 mL
Quantity 1 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.