Learn More
Thermo Scientific™ HyperSep™ SCX Strong Cation Exchanger SPE 96-Well Plates and Individual Wells
Catalog No. 03251210 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep SCX
Catalog No. Sorbent
03-251-210 HyperSep SCX
1 options
Catalog No. 03-251-210 Supplier Thermo Scientific™ Supplier No. 60300582
Add to Cart
Add to Cart
Obtain excellent performance for extraction of charged basic compounds with these strong cation exchanger SPE well plates and individual wells.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep SCX Strong Cation Exchanger SPE 96-Well Plates and Individual Wells feature a strong cation exchange sorbent for extraction of charged basic compounds. Typical applications include extraction of:
Antibiotics
Drug compounds
Organic bases
Amino acids
Catecholamines
Herbicides
HyperSep SCX Strong Cation Exchanger 96-Well Plates and Individual Wells are ideally suited for the extraction of a wide range of charged basic compounds.
Modified benzene ring provides potential for nonpolar interactions
Low pKa value
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Description Individual SPE wells; Bed weight 25 mg
For Use With (Application) Extraction of Charged Basic Compounds
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep SCX
Type Individual Well