Learn More
Thermo Scientific™ HyperSep™ SPE Plates
Catalog No. 03150548
Change view
Click to view available options
Includes:
10 mg/1 mL 96-well plate
20 mg/1 mL 96-well plate
3 mg/0.6 mL 96-well plate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
03-150-548 10 mg/1 mL 96-well plate
03-150-551 20 mg/1 mL 96-well plate
03-150-550 3 mg/0.6 mL 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-150-548 Supplier Thermo Scientific™ Supplier No. 6030010196WAX
Only null left
Add to Cart
Add to Cart
Innovative HyperSep™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ HyperSep™ Solid Phase Extraction Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, HyperSep SPE Well Plates deliver unparalleled performance, including:
  • Higher levels of reproducibility
  • Lower elution volumes
  • High extract cleanliness
HyperSep Solid Phase Extraction Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You'll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
  • 10 mg/2 mL 96 well plate format
  • Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
  • HyperSep products are also available in cartridge format.
TRUSTED_SUSTAINABILITY
Specifications
Bed Weight 10 mg
For Use With (Application) Bioanalytical Applications - Weak Ion-Exchange Retention of Strong Acidic Compounds. Sorbent Charge Can be Activated or Deactivated. Complementary Reversed Phase Retention of Neutral Compounds
Product Line HyperSep
Type 96-well Plate
Volume (Metric) Well 1 mL
Includes 10 mg/1 mL 96-well plate