Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Catalog No. 60104289 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
24 Position Extraction Plate Plugs
24 Well Extraction Plate
48 Well Extraction Plate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
60-104-289 48 Well Extraction Plate
03-251-255 24 Position Extraction Plate Plugs
60-104-284 24 Well Extraction Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-104-289 Supplier Thermo Scientific™ Supplier No. 60104289
Use these 24 or 48 well extraction plates and plugs with the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold.
Use these 24 or 48 well extraction plates and plugs with Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. This manifold process your samples in solid phase extraction SPE cartridges or 96 well plates.
TRUSTED_SUSTAINABILITY
Specifications
Description 48 Well Extraction Plate, 1 pk
For Use With (Application) SPE Sample Processing
For Use With (Equipment) Universal Vacuum Manifold
Product Line HyperSep
Type 48 Well Extraction Plate