Learn More
MilliporeSigma™ Supelco™ HyperShear Ultra HPLC High Pressure Static Mixer, Binary Tee Housing Assembly, 18,000 psi
Catalog No. 111007449 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
50 μL
150 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-100-7449 150 μL
11-100-7448 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-7449 Supplier MilliporeSigma™ Supelco™ Supplier No. 56649U
Add to Cart
Add to Cart
HyperShear Mixers incorporate a highly efficient cross-flow shearing mechanism which produces vortex shear mixing over a wide flow range. This mixing technology typically delivers between 25% to 200% better mixing efficiency compared to conventional packed bed or tortuous path mixers.
TRUSTED_SUSTAINABILITY
Specifications
Material 316 Stainless Steel/PEEK
Description Port size, 10-32 CPI (standard HPLC fittngs for 1/16 in. tubing)
Volume (Metric) 150 μL