Learn More
Thermo Scientific™ Hypersil GOLD™ C8 Reversed Phase HPLC Column, 5 μm, 4 mm x 125 mm
Column Format: Analytical Column; Carbon Load: 8%; Pore Size: 175 Å; Column Phase: Reversed Phase
Supplier:  Thermo Scientific™ 365D0321
Catalog No. 365D0321
Add to Cart
Description
Description
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions