Learn More
Med Vet International Hypertonic Saline 7.2% Sterile PreservatIVe Free Solution, 1000mL, Each
Catalog No. 502828505
Encompass_Preferred
Sterile, preservative-free hypertonic saline solution for use in replacement therapy of sodium, chloride and water, which may become depleted in many diseases. Each 100 mL contains 7.2 grams of sodium chloride. For use in horses and cattle.
Catalog No. 50-282-8505 Supplier Med Vet International Supplier No. RXHYPER7.21000V
Sterile, preservative-free hypertonic saline solution for use in replacement therapy of sodium, chloride and water, which may become depleted in many diseases. Each 100 mL contains 7.2 grams of sodium chloride. For use in horses and cattle.