Learn More
Apexbio Technology LLC HyperTrap AR rProtein A FF Column 5ml
SDP
Catalog No. 5000092914
Encompass_Preferred
HyperTrap AR rProtein A FF Column is a chromatography medium designed for effective purification of recombinant proteins particularly immunoglobulins through affinity-based interactions The column utilizes recombinant Protein A which selectively binds to the Fc region of antibodies enabling efficient capture and isolation from complex biological samples Its robust binding capacity and high flow rate support rapid processing in downstream bioprocessing and analytical workflows This product is suited for antibody purification contaminant removal and related research applications in protein chemistry and bioprocess development
Catalog No. 50-000-92914 Supplier Apexbio Technology LLC Supplier No. PC100551
HyperTrap AR rProtein A FF Column is a chromatography medium designed for effective purification of recombinant proteins particularly immunoglobulins through affinity-based interactions The column utilizes recombinant Protein A which selectively binds to the Fc region of antibodies enabling efficient capture and isolation from complex biological samples Its robust binding capacity and high flow rate support rapid processing in downstream bioprocessing and analytical workflows This product is suited for antibody purification contaminant removal and related research applications in protein chemistry and bioprocess development