Learn More
Apexbio Technology LLC HyperTrap Glutathione 4FF Column 5x5ml
SDP
Catalog No. 5000094895
Encompass_Preferred
The HyperTrap Glutathione 4FF Column is a specialized affinity chromatography medium designed for the selective binding and purification of glutathione S-transferase (GST)-tagged proteins and glutathione-reactive biomolecules The column utilizes immobilized glutathione on a highly cross-linked 4FF agarose matrix enabling efficient capture of GST fusion proteins through specific glutathione-protein interactions This technology supports downstream applications such as protein purification functional characterization and interaction studies in molecular biology and biochemistry research The column s high binding capacity and stability make it suitable for preparative and analytical workflows requiring selective protein isolation
Catalog No. 50-000-94895 Supplier Apexbio Technology LLC Supplier No. PC100325
The HyperTrap Glutathione 4FF Column is a specialized affinity chromatography medium designed for the selective binding and purification of glutathione S-transferase (GST)-tagged proteins and glutathione-reactive biomolecules The column utilizes immobilized glutathione on a highly cross-linked 4FF agarose matrix enabling efficient capture of GST fusion proteins through specific glutathione-protein interactions This technology supports downstream applications such as protein purification functional characterization and interaction studies in molecular biology and biochemistry research The column s high binding capacity and stability make it suitable for preparative and analytical workflows requiring selective protein isolation