Learn More
Apexbio Technology LLC HyperTrap Heparin FF Column 5x5ml
SDP
Catalog No. 5000094896
Encompass_Preferred
HyperTrap Heparin FF Column is an affinity chromatography medium designed for the purification of heparin-binding biomolecules The column exploits the strong ionic and specific interactions between immobilized heparin and target proteins such as growth factors coagulation factors and certain enzymes enabling selective retention and efficient separation from complex biological mixtures This tool is widely utilized in the isolation and characterization of heparin-binding proteins investigation of protein glycosaminoglycan interactions and purification of bioactive factors for downstream functional assays and structural analyses
Catalog No. 50-000-94896 Supplier Apexbio Technology LLC Supplier No. PC100825
HyperTrap Heparin FF Column is an affinity chromatography medium designed for the purification of heparin-binding biomolecules The column exploits the strong ionic and specific interactions between immobilized heparin and target proteins such as growth factors coagulation factors and certain enzymes enabling selective retention and efficient separation from complex biological mixtures This tool is widely utilized in the isolation and characterization of heparin-binding proteins investigation of protein glycosaminoglycan interactions and purification of bioactive factors for downstream functional assays and structural analyses