Learn More
Apexbio Technology LLC HyperTrap-Phenyl-6FF-HS-Column
SDP
Catalog No. 5000570790
Encompass_Preferred
HyperTrap Phenyl 6FF (HS) Column is a specialized tool used in biochemical research particularly in chromatographic applications for protein purification and separation Originating from advanced polymer technology it utilizes a hydrophobic interaction mechanism where phenyl groups interact with target molecules facilitating selective affinity and binding This column is essential for exploring protein-protein interactions enzyme assays and biomarker identification demonstrating activity in varying ranges predominantly influenced by specific experimental setups Commonly applied in cell-based assays animal model studies and high-throughput drug screening it supports biomolecular research across diverse disciplines Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70790 Supplier Apexbio Technology LLC Supplier No. PC10185
HyperTrap Phenyl 6FF (HS) Column is a specialized tool used in biochemical research particularly in chromatographic applications for protein purification and separation Originating from advanced polymer technology it utilizes a hydrophobic interaction mechanism where phenyl groups interact with target molecules facilitating selective affinity and binding This column is essential for exploring protein-protein interactions enzyme assays and biomarker identification demonstrating activity in varying ranges predominantly influenced by specific experimental setups Commonly applied in cell-based assays animal model studies and high-throughput drug screening it supports biomolecular research across diverse disciplines Experimental concentrations typically depend on the specific research design