Learn More
Apexbio Technology LLC HYPERTRAP-PHENYL-HP-COLUMN-5ML
SDP
Catalog No. 5000570759
Encompass_Preferred
HyperTrap Phenyl HP Column is a specialized tool used in the biomedical research field particularly in chromatography applications for the separation and analysis of complex mixtures Originating from advanced materials science this column utilizes phenyl-functionalized silica particles to enhance its selectivity and binding affinity in various chromatographic processes The mechanism of action focuses on interactions between the phenyl groups and target molecules often used to study pathways involving hydrophobic and - interactions In vitro activity typically ranges from nanomolar to low micromolar concentrations enabling precise analysis and separation in a variety of settings This column is widely employed in biochemical research including cell model studies animal studies and drug screening processes Experimental concentrations typically depend on the specific research design ensuring flexibility and adaptability to diverse experimental needs
Catalog No. 50-005-70759 Supplier Apexbio Technology LLC Supplier No. PC101951
Add to Cart
Add to Cart
HyperTrap Phenyl HP Column is a specialized tool used in the biomedical research field particularly in chromatography applications for the separation and analysis of complex mixtures Originating from advanced materials science this column utilizes phenyl-functionalized silica particles to enhance its selectivity and binding affinity in various chromatographic processes The mechanism of action focuses on interactions between the phenyl groups and target molecules often used to study pathways involving hydrophobic and - interactions In vitro activity typically ranges from nanomolar to low micromolar concentrations enabling precise analysis and separation in a variety of settings This column is widely employed in biochemical research including cell model studies animal studies and drug screening processes Experimental concentrations typically depend on the specific research design ensuring flexibility and adaptability to diverse experimental needs