Learn More
Apexbio Technology LLC HyperTrap Protein G 4FF Column 1ml
SDP
Catalog No. 5000094835
Encompass_Preferred
HyperTrap Protein G 4FF Column is an affinity chromatography medium designed for the purification of immunoglobulins particularly IgG from complex biological samples The column features immobilized Protein G ligands which specifically bind to the Fc region of IgG molecules via non-covalent interactions allowing selective capture and efficient separation of antibodies This mechanism facilitates high-purity isolation of immunoglobulins without the need for harsh elution conditions The HyperTrap Protein G 4FF Column is widely utilized in biomedical research for antibody purification immunoprecipitation and preparative workflows requiring reproducible antibody isolation
Catalog No. 50-000-94835 Supplier Apexbio Technology LLC Supplier No. PC10061
HyperTrap Protein G 4FF Column is an affinity chromatography medium designed for the purification of immunoglobulins particularly IgG from complex biological samples The column features immobilized Protein G ligands which specifically bind to the Fc region of IgG molecules via non-covalent interactions allowing selective capture and efficient separation of antibodies This mechanism facilitates high-purity isolation of immunoglobulins without the need for harsh elution conditions The HyperTrap Protein G 4FF Column is widely utilized in biomedical research for antibody purification immunoprecipitation and preparative workflows requiring reproducible antibody isolation