Learn More
Apexbio Technology LLC HyperTrap Q FF Column 1ml
SDP
Catalog No. 5000092912
Encompass_Preferred
The HyperTrap Q FF Column is not a small molecule but a chromatography medium designed for the purification of biomolecules particularly proteins and nucleic acids It operates via strong anion exchange chromatography utilizing quaternary ammonium functional groups immobilized on a Fast Flow resin matrix to selectively bind negatively charged molecules under defined buffer conditions This allows for efficient separation and recovery of target biomolecules from complex mixtures The HyperTrap Q FF Column is applied in research settings for desalting buffer exchange and isolation of nucleic acids or proteins supporting downstream analyses and preparative workflows in the biomedical field
Catalog No. 50-000-92912 Supplier Apexbio Technology LLC Supplier No. PC10101
The HyperTrap Q FF Column is not a small molecule but a chromatography medium designed for the purification of biomolecules particularly proteins and nucleic acids It operates via strong anion exchange chromatography utilizing quaternary ammonium functional groups immobilized on a Fast Flow resin matrix to selectively bind negatively charged molecules under defined buffer conditions This allows for efficient separation and recovery of target biomolecules from complex mixtures The HyperTrap Q FF Column is applied in research settings for desalting buffer exchange and isolation of nucleic acids or proteins supporting downstream analyses and preparative workflows in the biomedical field