Learn More
Apexbio Technology LLC HyperTrap Q HP Column 1ml
SDP
Catalog No. 5000092910
Encompass_Preferred
HyperTrap Q HP Column is not a small molecule but a high-performance chromatography column designed for biomolecule purification Featuring a strong anion exchange resin with quaternary ammonium functional groups the column enables efficient separation and capture of negatively charged biomolecules such as proteins and nucleic acids based on their charge properties The column supports high binding capacity and robust performance across a range of pH conditions making it suitable for process development scale-up and routine purification workflows in biomedical research particularly for the preparation and analysis of protein therapeutics and other biologically relevant molecules
Catalog No. 50-000-92910 Supplier Apexbio Technology LLC Supplier No. PC10111
HyperTrap Q HP Column is not a small molecule but a high-performance chromatography column designed for biomolecule purification Featuring a strong anion exchange resin with quaternary ammonium functional groups the column enables efficient separation and capture of negatively charged biomolecules such as proteins and nucleic acids based on their charge properties The column supports high binding capacity and robust performance across a range of pH conditions making it suitable for process development scale-up and routine purification workflows in biomedical research particularly for the preparation and analysis of protein therapeutics and other biologically relevant molecules