Learn More
Apexbio Technology LLC HyperTrap SP HP Column 5ml
SDP
Supplier:  Apexbio Technology LLC PC101551
Encompass_Preferred
HyperTrap SP HP Column is a chromatography medium designed for the efficient purification of biomolecules based on strong cation exchange principles The column utilizes sulfopropyl (SP) ligands immobilized on a high-performance matrix allowing selective binding of positively charged proteins and peptides under controlled buffer conditions This mechanism facilitates the separation and enrichment of target analytes from complex biological samples The HyperTrap SP HP Column is commonly applied in protein purification workflows biomarker discovery and quality control assays within biomedical research
Catalog No. 50-000-94854