Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hypidone hydrochloride | 1339058-04-6 | 99.7% | 334.84 g/mol | C18H23ClN2O2 | 1 ML
SDP

Catalog No. 5000213552
Encompass_Preferred

Hypidone hydrochloride is a research-grade small molecule reported to act as an orally active antidepressant agent with activity at serotonin (5-HT) receptors. It is provided for laboratory research use and is available both as solid quantities and as a ready-to-use solution in DMSO, with supporting product documentation and safety data.

  • Research-grade 5-HT receptor agonist for laboratory studies.
  • High purity suitable for research (reported 99.73%).
  • Available as solids and as a 10 mM, 1 mL solution in DMSO.
  • Chemical identifiers: CAS 1339058-04-6; formula C18H23ClN2O2; MW 334.84 g/mol.
  • Intended for in vitro and in vivo research; not for human use.
  • Supplied with datasheet and safety data sheet for handling and storage.

Catalog No. 50-002-13552 Supplier Medchemexpress LLC Supplier No. HY10076910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hypidone hydrochloride is a research-grade small molecule reported to act as an orally active antidepressant agent with activity at serotonin (5-HT) receptors. It is provided for laboratory research use and is available both as solid quantities and as a ready-to-use solution in DMSO, with supporting product documentation and safety data.

  • Research-grade 5-HT receptor agonist for laboratory studies.
  • High purity suitable for research (reported 99.73%).
  • Available as solids and as a 10 mM, 1 mL solution in DMSO.
  • Chemical identifiers: CAS 1339058-04-6; formula C18H23ClN2O2; MW 334.84 g/mol.
  • Intended for in vitro and in vivo research; not for human use.
  • Supplied with datasheet and safety data sheet for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.