Learn More
Medchemexpress LLC Hypidone hydrochloride | 1339058-04-6 | 99.7% | 334.84 g/mol | C18H23ClN2O2 | 10 MG
SDP
Catalog No. 5000213129
Encompass_Preferred
Hypidone hydrochloride is the hydrochloride salt of a research compound reported to modulate 5-HT receptors and inhibit serotonin uptake in preclinical studies. It is supplied as a high-purity solid for laboratory use, typically in milligram-scale vials for in vitro and in vivo research.
  • High reported purity (>99%).
  • Molecular weight 334.84 g/mol.
  • Solid hydrochloride salt, white to off-white powder.
  • Reported activity as a 5-HT receptor modulator and uptake inhibitor in preclinical models.
  • Packaged in small, milligram-scale quantities suitable for research.
Catalog No. 50-002-13129 Supplier Medchemexpress LLC Supplier No. HY10076910MG
Hypidone hydrochloride is the hydrochloride salt of a research compound reported to modulate 5-HT receptors and inhibit serotonin uptake in preclinical studies. It is supplied as a high-purity solid for laboratory use, typically in milligram-scale vials for in vitro and in vivo research.
  • High reported purity (>99%).
  • Molecular weight 334.84 g/mol.
  • Solid hydrochloride salt, white to off-white powder.
  • Reported activity as a 5-HT receptor modulator and uptake inhibitor in preclinical models.
  • Packaged in small, milligram-scale quantities suitable for research.