Learn More
Medchemexpress LLC Hypidone hydrochloride | 1339058-04-6 | 99.7% | 334.84 g/mol | C18H23ClN2O2 | 100 MG
SDP
Catalog No. 5000211328
Encompass_Preferred
Hypidone hydrochloride is the hydrochloride salt of a research compound with potent serotonergic activity. It functions as a selective serotonin (5-HT) uptake inhibitor and a 5-HT1A receptor agonist, and has demonstrated oral activity in animal models of depression. Supplied as a powder for laboratory research use only; not for human or clinical use.
  • High purity suitable for research applications.
  • Dual mechanism: serotonin uptake inhibition and 5-HT1A receptor agonism.
  • Demonstrated oral activity in animal depression models.
  • Available in multiple pack sizes including small research quantities.
  • Recommended sealed storage away from moisture; in solvent store at -80°C for long term.
Catalog No. 50-002-11328 Supplier Medchemexpress LLC Supplier No. HY100769100MG
Hypidone hydrochloride is the hydrochloride salt of a research compound with potent serotonergic activity. It functions as a selective serotonin (5-HT) uptake inhibitor and a 5-HT1A receptor agonist, and has demonstrated oral activity in animal models of depression. Supplied as a powder for laboratory research use only; not for human or clinical use.
  • High purity suitable for research applications.
  • Dual mechanism: serotonin uptake inhibition and 5-HT1A receptor agonism.
  • Demonstrated oral activity in animal depression models.
  • Available in multiple pack sizes including small research quantities.
  • Recommended sealed storage away from moisture; in solvent store at -80°C for long term.