Learn More
Medchemexpress LLC Hypidone hydrochloride | 1339058-04-6 | 99.7% | 334.84 g/mol | C18H23ClN2O2 | 5 MG
SDP
Catalog No. 5000211862
Encompass_Preferred
Hypidone hydrochloride is a research-grade, orally active antidepressant used in preclinical pharmacology to study serotonergic mechanisms. It functions as a selective serotonin uptake inhibitor and a potent 5-HT1A receptor agonist, showing low-nanomolar activity in uptake and receptor assays. Supplied as a white to off-white solid with high purity for laboratory use.
  • Orally active antidepressant with dual mechanism of action.
  • Highly potent 5-HT uptake inhibitor and 5-HT1A receptor agonist.
  • High purity suitable for preclinical research.
  • Available in small milligram package sizes and DMSO solution format.
  • Soluble in DMSO and supplied with recommended storage conditions.
Catalog No. 50-002-11862 Supplier Medchemexpress LLC Supplier No. HY1007695MG
Hypidone hydrochloride is a research-grade, orally active antidepressant used in preclinical pharmacology to study serotonergic mechanisms. It functions as a selective serotonin uptake inhibitor and a potent 5-HT1A receptor agonist, showing low-nanomolar activity in uptake and receptor assays. Supplied as a white to off-white solid with high purity for laboratory use.
  • Orally active antidepressant with dual mechanism of action.
  • Highly potent 5-HT uptake inhibitor and 5-HT1A receptor agonist.
  • High purity suitable for preclinical research.
  • Available in small milligram package sizes and DMSO solution format.
  • Soluble in DMSO and supplied with recommended storage conditions.