Learn More
Medchemexpress LLC Hypidone hydrochloride | 1339058-04-6 | 99.7% | 334.84 g/mol | C18H23ClN2O2 | 50 MG
SDP
Catalog No. 5000211863
Encompass_Preferred
Hypidone hydrochloride (YL0919) is a research-stage small molecule characterized as a 5-HT (serotonin) receptor agonist. Supplied as the hydrochloride salt, it is intended for in vitro neuroscience and pharmacology research and is provided with supporting analytical documentation.
  • Characterized as a 5-HT receptor agonist for neuroscience research.
  • Supplied as the hydrochloride salt suitable for laboratory use.
  • High purity (99.7%) as reported by the manufacturer.
  • Available in small-mass formats and as a 10 mM solution in DMSO.
  • Provided with datasheet, SDS, COA, and LCMS documentation.
Catalog No. 50-002-11863 Supplier Medchemexpress LLC Supplier No. HY10076950MG
Hypidone hydrochloride (YL0919) is a research-stage small molecule characterized as a 5-HT (serotonin) receptor agonist. Supplied as the hydrochloride salt, it is intended for in vitro neuroscience and pharmacology research and is provided with supporting analytical documentation.
  • Characterized as a 5-HT receptor agonist for neuroscience research.
  • Supplied as the hydrochloride salt suitable for laboratory use.
  • High purity (99.7%) as reported by the manufacturer.
  • Available in small-mass formats and as a 10 mM solution in DMSO.
  • Provided with datasheet, SDS, COA, and LCMS documentation.