Learn More
Thermo Scientific™ Hypo-Vial Septa
Catalog No. PI13237 Shop All Thermo Scientific Products
Catalog No. PI13237 Supplier Thermo Scientific™ Supplier No. TS13237
Thermo Scientific™ Hypo-Vial Septa are ideal to use with borosilicate glass sample bottles for heating and autoclaving.
Thermo Scientific™ Hypo-Vial Septa are ideal to use with borosilicate glass sample bottles for heating and autoclaving.
  • Closure for 50 and 125mL glass sample bottles
  • Hycar and silicone plug-type septa and flat PTFE/Silicone laminated discs (sold separately)
  • Fits 6-125mL Hypo-Vial sample storage vials
  • Enable syringe-needle access to vial contents
TRUSTED_SUSTAINABILITY
Specifications
Description Silicone Septa
Material Silicone
For Use With (Equipment) Borosilicate Glass Sample Bottles for Heating and Autoclaving
Product Type For 6-125 mL Hypo-Vial Sample Storage Vials