Learn More
Pickering Laboratories Inc HYPOCHLORITE DILUENT 4X950ML
SDP
Catalog No. NC9200912
Encompass_Preferred
Hypochlorite Diluent for Glyphosate analysis, case of 4 (950 mL/bottle) EPA 547
Catalog No. NC9200912 Supplier Pickering Laboratories Inc Supplier No. GA116
Hypochlorite Diluent for Glyphosate analysis, case of 4 (950 mL/bottle) EPA 547