Learn More
Cayman Chemical Hypocrellin A, Purity: ≥98%, CAS: 77029-83-5, 10 mg
SDP
Catalog No. 502988911
Encompass_Preferred
A perylenequinoid with diverse biological activities; active against C. albicans, C. parapsilosis, and C. tropicalis fungi (IC50s = 0.65, 1, and 85 µg/ml, respectively), as well as S. aureus, MRSA, P. aeruginosa, and M. intracellulare bacteria (IC50s = 3, 7, 10, and 2.5 µg/ml, respectively); active against L. donovani promastigotes (IC50 = 0.27 µg/ml); induces production of reactive oxygen species and apoptosis and reduces cell viability of A549 cancer cells upon photoactivation with UV radiation
Catalog No. 50-298-8911 Supplier Cayman Chemical Supplier No. 2993310 MG
A perylenequinoid with diverse biological activities; active against C. albicans, C. parapsilosis, and C. tropicalis fungi (IC50s = 0.65, 1, and 85 µg/ml, respectively), as well as S. aureus, MRSA, P. aeruginosa, and M. intracellulare bacteria (IC50s = 3, 7, 10, and 2.5 µg/ml, respectively); active against L. donovani promastigotes (IC50 = 0.27 µg/ml); induces production of reactive oxygen species and apoptosis and reduces cell viability of A549 cancer cells upon photoactivation with UV radiation