Learn More
Cayman Chemical HypocrelIn B 10mg
SDP
Catalog No. 502940778
Encompass_Preferred
A fungal metabolite; decreases the viability of, and induces apoptosis in, MDA-MB-231 breast cancer cells when used in the presence of ultrasound exposure; increases the production of ROS in MDA-MB-231 cells at 2.5 µM with concomitant ultrasound exposure; active against MRSA at 20 µM in combination with ultrasound sonication
Catalog No. 50-294-0778 Supplier Cayman Chemical Supplier No. 2854410 MG
A fungal metabolite; decreases the viability of, and induces apoptosis in, MDA-MB-231 breast cancer cells when used in the presence of ultrasound exposure; increases the production of ROS in MDA-MB-231 cells at 2.5 µM with concomitant ultrasound exposure; active against MRSA at 20 µM in combination with ultrasound sonication