Learn More
Medchemexpress LLC Hypocrellin B | 123940-54-5 | 528.51 | 100 MG
SDP
Catalog No. 5000415178
Encompass_Preferred
Hypocrellin B is a pigment isolated from the fungi Hypocrella bambusae and Shiraia bambusicola. It is an apoptosis inducer and can be used as a photosensitizer for photodynamic therapy of cancer. It also exhibits antimicrobial and antileishmanial activities.
  • Apoptosis inducer
  • Photosensitizer for cancer photodynamic therapy
  • Exhibits antimicrobial activity
  • Exhibits antileishmanial activity
  • Cytotoxic against BGC-823 cells (IC50: 40 nM)
  • Induces early apoptosis in HepG2 cells
  • Antifungal activity against C. albicans (IC50: 5 μg/mL)
  • Anti-parasite activity against Leishmania donovani (IC50: 12.7 μg/mL)
Catalog No. 50-004-15178 Supplier Medchemexpress LLC Supplier No. HYN1453100MG
Hypocrellin B is a pigment isolated from the fungi Hypocrella bambusae and Shiraia bambusicola. It is an apoptosis inducer and can be used as a photosensitizer for photodynamic therapy of cancer. It also exhibits antimicrobial and antileishmanial activities.
  • Apoptosis inducer
  • Photosensitizer for cancer photodynamic therapy
  • Exhibits antimicrobial activity
  • Exhibits antileishmanial activity
  • Cytotoxic against BGC-823 cells (IC50: 40 nM)
  • Induces early apoptosis in HepG2 cells
  • Antifungal activity against C. albicans (IC50: 5 μg/mL)
  • Anti-parasite activity against Leishmania donovani (IC50: 12.7 μg/mL)