Learn More
Medchemexpress LLC Hypocrellin B | 123940-54-5 | 528.51 | 25 MG
SDP
Catalog No. 5000390314
Encompass_Preferred
Hypocrellin B is a pigment isolated from the fungi Hypocrella bambusae and Shiraia bambusicola. It acts as an apoptosis inducer and can be used as a photosensitizer for photodynamic therapy of cancer. It also possesses antimicrobial and antileishmanial activities.
  • Apoptosis inducer
  • Photosensitizer for photodynamic therapy of cancer
  • Antimicrobial activity
  • Antileishmanial activity
  • Induces cell cytotoxicity and early apoptosis in HepG2 cells with sonodynamic therapy
  • Shows antifungal activity against C. albicans
  • Shows anti-parasite activity against Leishmania donovani
  • Inhibits tumor growth in A549 tumor bearing mice
Catalog No. 50-003-90314 Supplier Medchemexpress LLC Supplier No. 25MGHYN1453
Hypocrellin B is a pigment isolated from the fungi Hypocrella bambusae and Shiraia bambusicola. It acts as an apoptosis inducer and can be used as a photosensitizer for photodynamic therapy of cancer. It also possesses antimicrobial and antileishmanial activities.
  • Apoptosis inducer
  • Photosensitizer for photodynamic therapy of cancer
  • Antimicrobial activity
  • Antileishmanial activity
  • Induces cell cytotoxicity and early apoptosis in HepG2 cells with sonodynamic therapy
  • Shows antifungal activity against C. albicans
  • Shows anti-parasite activity against Leishmania donovani
  • Inhibits tumor growth in A549 tumor bearing mice