Learn More
TARGETMOL CHEMICALS INC HYPOCRELLIN B 5MG
SDP
Catalog No. 502876779
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. HYPOCRELLIN B are photosensitive pigments isolated from Hypocrella bambusae Sacc. Hypocrellin B causes DNA strand breakage, induces apoptosis in ovarian cancer cells, and inhibits proliferation of Staphylococcus by increasing ROS levels, and damaging cell walls. Purity 98.87%
Catalog No. 50-287-6779 Supplier TARGETMOL CHEMICALS INC Supplier No. T57805MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. HYPOCRELLIN B are photosensitive pigments isolated from Hypocrella bambusae Sacc. Hypocrellin B causes DNA strand breakage, induces apoptosis in ovarian cancer cells, and inhibits proliferation of Staphylococcus by increasing ROS levels, and damaging cell walls. Purity 98.87%