Learn More
Medchemexpress LLC HY-N1453 5mg Medchemexpress, Hypocrellin B CAS:123940-54-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN14535MG
Encompass_Preferred
Medchemexpress, HY-N1453 5mg Hypocrellin B CAS:123940-54-5 Hypocrellin B, a pigment isolated from the fungi Hypocrella bambusae and Shiraia bambusicola, is an apoptosis inducer. Hypocrellin B can be used as a photosensitizer for photodynamic therapy of cancer. Hypocrellin B also has antimicrobial and antileishmanial activities[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0342