Learn More
Exel Corporation Hypodermic Needle Specialty Use
Catalog No. 22355129
Change view
Click to view available options
Needle Gauge:
31 G
32 G
33 G
Color:
Clear
Green
Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
22-355-129 33 G Clear
22-355-126 31 G Orange
22-355-127 32 G Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-355-129 Supplier Exel Corporation Supplier No. EXE 26393
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Hypodermic Needle Specialty Use is designed with thin walls to deliver more viscous fluids. The needles are manufactured with SUS - 304 Stainless Steel. A variety of sizes are available. Available in bulk.
  • Sterile, Non-Toxic, Non-Pyrogenic
  • SUS – 304 Stainless Steel
  • Needles and boxes are color-coded for easy identification
  • Each needle is individually packaged, sterilized and ready for use
  • This product does not contain Natural Rubber Latex
Specifications
Material Stainless Steel (304)
Color Clear
Needle Gauge 33 G
Latex Free Yes
Length (English) Needle 1⁄2 in.
Product Type Hypodermic Needle
Quantity 2000/Cs.
Sterility Sterile