Learn More
Exel Corporation Hypodermic Needle Specialty Use Soft Pack
Catalog No. 22355128
Change view
Click to view available options
Needle Gauge:
32 G
Catalog No. Needle Gauge
22-355-128 32 G
1 options
This item is not returnable. View return policy
Catalog No. 22-355-128 Supplier Exel Corporation Supplier No. EXE 26392S
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Hypodermic Needle Specialty Use is designed with thin walls to deliver more viscous fluids. The needles are manufactured with SUS - 304 Stainless Steel. A variety of sizes are available. Available in bulk.
  • Sterile, Non-Toxic, Non-Pyrogenic
  • SUS – 304 Stainless Steel
  • Needles and boxes are color-coded for easy identification
  • Each needle is individually packaged, sterilized and ready for use
  • This product does not contain Natural Rubber Latex
  • Soft Pack
Specifications
Material Stainless Steel (304)
Color Green
Needle Gauge 32 G
Latex Free Yes
Length (English) Needle 1⁄2 in.
Product Type Hypodermic Needle
Packaging Soft Pk.
Sterility Sterile