Learn More
Medchemexpress LLC α-Hypophamine (acetate); Oxytocic hormone (acetate) | 6233-83-6 | 99.7% | 1067.24 | 25 MG
SDP
Catalog No. 5000383956
Encompass_Preferred
Oxytocin (α-Hypophamine) acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. It can also function as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects, particularly in the face of adversity or trauma.
  • Facilitates parturition, lactation, and prosocial behaviors.
  • Functions as a stress-coping molecule.
  • Exhibits anti-inflammatory effects.
  • Possesses antioxidant properties.
  • Provides protective effects, especially against adversity or trauma.
Catalog No. 50-003-83956 Supplier Medchemexpress LLC Supplier No. 25MGHY17571A
Oxytocin (α-Hypophamine) acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. It can also function as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects, particularly in the face of adversity or trauma.
  • Facilitates parturition, lactation, and prosocial behaviors.
  • Functions as a stress-coping molecule.
  • Exhibits anti-inflammatory effects.
  • Possesses antioxidant properties.
  • Provides protective effects, especially against adversity or trauma.