Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypophosphorous acid, 50% w/w aq. soln.

Catalog No. AA1414257
Change view
Click to view available options
4x500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA1414257 4x500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1414257 Supplier Thermo Scientific™ Supplier No. 1414257
Request Bulk or Custom Formats

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

CAS 6303-21-5
Density 1.225
Quantity 4 x 500 g
Molecular Formula H{3}O{2}P
Linear Formula H{3}PO{2}
Physical Form Liquid
MDL Number MFCD02183592
UN Number UN3264
Synonym Phosphinic acid;
Formula Weight 66.00
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.