Learn More
Sigma Aldrich Fine Chemicals Biosciences Hypophosphorous acid solut4KG
Catalog No. 502902986
Encompass
H3PO2 a monobasic oxyacid is also referred to as phosphinic acid. H3PO2 participates as a reducing agent for the conversion of aromatic and aliphatic diselenides to the corresponding selenols.It undergoes oxidation in the presence of Ce(IV) and a strong acid to afford cerium(IV) hypophosphite complex ions.
Catalog No. 50-290-2986 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2149064X4KG
H3PO2 a monobasic oxyacid is also referred to as phosphinic acid. H3PO2 participates as a reducing agent for the conversion of aromatic and aliphatic diselenides to the corresponding selenols.It undergoes oxidation in the presence of Ce(IV) and a strong acid to afford cerium(IV) hypophosphite complex ions.