Learn More
Cayman Chemical Hypophyllanthin, Purity: ≥98%, CAS: 33676-00-5, 10 mg
SDP
Catalog No. 502953207
Encompass_Preferred
An active principle of P. amarus that has been shown to reversibly inhibit P-glycoprotein (also known as multidrug resistance protein 1 (MDR1)) function without effect on MDR2 activity
Catalog No. 50-295-3207 Supplier Cayman Chemical Supplier No. 1171510 MG
An active principle of P. amarus that has been shown to reversibly inhibit P-glycoprotein (also known as multidrug resistance protein 1 (MDR1)) function without effect on MDR2 activity