Learn More
Cayman Chemical Hypotaurine, Purity: ≥95%, CAS: 300-84-5, 100 mg
SDP
Catalog No. 502988630
Encompass_Preferred
An Eendogenous inhibitory amino acid; inhibits GABA uptake by mouse GAT1, GAT2, GAT3, and GAT4 (IC50s = 170, 240, 4.9, and 8.1 μM, respectively) and rat GAT1, GAT2, and GAT3 (IC50s = 1,010, 52, and 73 μM, respectively); inhibits GABA uptake by BGT1 n L-M(TK-) cells expressing the human transporter (IC50 = 380 μM); inhibits sodium-stimulated GABA uptake by rabbit choroid plexus slices (IC50 = 21.9 μM); scavenges HOCl and hydroxyl, but not superoxide, radicals in cell-free assays; protects rat TR-TBT 18d-1 placental trophoblasts from hydrogen peroxide-induced cell death at 1 mM
Catalog No. 50-298-8630 Supplier Cayman Chemical Supplier No. 29614100 MG
An Eendogenous inhibitory amino acid; inhibits GABA uptake by mouse GAT1, GAT2, GAT3, and GAT4 (IC50s = 170, 240, 4.9, and 8.1 μM, respectively) and rat GAT1, GAT2, and GAT3 (IC50s = 1,010, 52, and 73 μM, respectively); inhibits GABA uptake by BGT1 n L-M(TK-) cells expressing the human transporter (IC50 = 380 μM); inhibits sodium-stimulated GABA uptake by rabbit choroid plexus slices (IC50 = 21.9 μM); scavenges HOCl and hydroxyl, but not superoxide, radicals in cell-free assays; protects rat TR-TBT 18d-1 placental trophoblasts from hydrogen peroxide-induced cell death at 1 mM