Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hypotaurine

Catalog No. 5000015026 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-000-15026 100 mg
H1384 250 mg
H138410MG 10 mg
50-000-15027 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-000-15026 Supplier Sigma Aldrich Supplier No. H1384100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Hypotaurine has been used as a supplement in fertilization medium (fert- tyrode albumin lactate pyruvate (talp)), which used in in-vitro fertilization and evaluation of embryo development
  • It is also used as a h2s scavenger.hypotaurine, a cystamine analog, is the precursor for taurine synthesis
  • It can act as an antioxidant and singlet oxygen scavenger, thereby protecting cells from oxidative damage
  • Hypotaurine is reported to play an important role in the maintenance and sttandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • H1384.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 300-84-5
Molecular Formula C2H7NO2S
Linear Formula C2H7NO2S
MDL Number MFCD00038197
Quantity 100 mg
Synonym 2-Aminoethanesulfinic acid
Molecular Weight (g/mol) 109.15
Percent Purity ≥98% (TLC)
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.